EMAIL THIS PAGE TO A FRIEND
Comptes rendus des seances de la Societe de biologie et de ses filiales
[Inductive effects of enzymes of salicylic acid derivatives].
PMID 6220776
Abstract
In the series of salicylic acids derivatives investigated 3,5-DIS has a large inductive effect of enzymes with respect to the metabolism of xenobiotics: rise in the rate of Cyt. P 450; increase of ethoxycoumarin deethylase, bilirubin glucuronosyl transferase and benzphetamine-N-demethylase activities. This effect is comparable with that of phenobarbital.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
716405
2-Hydroxy-4-iodobenzoic acid, 97%
C7H5IO3