EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact allergy to N-hydroxyphthalimide.
PMID 6220864
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H53704
N-Hydroxyphthalimide, 97%
C8H5NO3