EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis from an organic di-isocyanate.
PMID 6225609
Abstract
Organic di-isocyanates are used widely in industry. The concomitant respiratory complications are well-known. Cutaneous problems precipitated by skin contact with these chemicals are less well recognised. Allergic contact dermatitis caused by exposure to di-cyclohexylmethane 4,4'-di-isocyanate has been demonstrated in a factory where it was used to form a surface-coating polyurethane. Attention to hygiene methods may have reduced the incidence of cases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388386
4,4′-Methylenebis(cyclohexyl isocyanate), mixture of isomers, 90%
C15H22N2O2