EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Activation of antitumor agent gilvocarcins by visible light.
PMID 6229029
Abstract
Gilvocarcins that are antitumor agents are activated by low doses of visible light to induce bacteriophage lambda in Escherichia coli. This result is dependent on interaction with DNA. Gilvocarcin M, an analog without antitumor activity, failed to induce the prophage after light exposure, thus demonstrating a correlation between photosensitizing and antitumor activities. These results raise several possibilities regarding the mode of action of gilvocarcins as antitumor agents in vivo, involving light or enzymatic activating systems, which could be exploited in human cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6616
Chrysomycin A, ≥98% (HPLC), solid
C28H28O9