EMAIL THIS PAGE TO A FRIEND
Zhonghua fang she xue za zhi Chinese journal of radiology
[Evaluation of 113mIn-DTPMP and 99mTc-MDP for bone scanning].
PMID 6236062
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36818
Diethylenetriaminepentakis(methylphosphonic acid) solution, technical, ~50% (T)
C9H28N3O15P5