EMAIL THIS PAGE TO A FRIEND
Anatomia, histologia, embryologia
Effect of isocyanide on the ultrastructure of mouse pineal.
PMID 6236716
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
133280
Butyl isocyanide, 97%
C5H9N