EMAIL THIS PAGE TO A FRIEND
Voprosy meditsinskoi khimii
[Changes in the secondary and tertiary structure of serum albumin in interactions with ligands of various structures].
PMID 6239449
Abstract
High affinity interactions between blood serum albumin and five substances of various chemical structure, exhibiting distinct physiological activity, were accompanied by alterations in the protein tertiary structure, while the albumin secondary structure was involved in conformational transformation after less effective affinity binding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45451
Dinobuton, PESTANAL®, analytical standard
C14H18N2O7