EMAIL THIS PAGE TO A FRIEND
Journal of steroid biochemistry
Effect of catechol estrogens and estriol on the induction of uterine peroxidase.
PMID 6247576
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H5517
2-Hydroxyestriol, ≥90% (HPLC), powder
C18H24O4