EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
The use of photolabels to probe the ouabain binding site of the (Na, K)-ATPase.
PMID 6247954
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30030
Cymarin, ≥96% (HPLC)
C30H44O9