EMAIL THIS PAGE TO A FRIEND
International journal of nuclear medicine and biology
The use of formamidine sulphinic acid in the preparation of 99mTc-labelled radiopharmaceuticals--a cautionary note.
PMID 6262265
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F16001
Formamidinesulfinic acid, ≥98%
CH4N2O2S