EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
Subsensitivity of the beta-adrenergic receptor-linked adenylate cyclase system of rat pineal gland following repeated treatment with desmethylimipramine and nialamide.
PMID 6262613
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
252999
Nialamide, 95%
C16H18N4O2