EMAIL THIS PAGE TO A FRIEND
Comptes rendus des seances de l'Academie des sciences. Serie III, Sciences de la vie
[Evidence, in vitro, of estrone and estradiol-17 beta 3-sulfoconjugate hydrolysis in the uterus of pregnant guinea pigs].
PMID 6263512
Abstract
The metabolism, in vitro, of [6,7-3H]-oestrone-3-sulfate and [6,7-3H]-oestradiol-17beta-3-sulfate by the uteri of pregnant female Guinea-Pigs has been studied using 900 g supernatants. A hydrolysis of the sulfate moiety was observed: 35% of the radioactivity was recovered in the unconjugated fractions after 1 hr. of incubation. Gluruconide oestrogens were also recovered but the rate of synthesis was very low (3.6% after 1 hr.). OEstrone and oestradiol-17beta have been identified in unconjugated, glucuro and sulfo-conjugated fractions after incubation with the tritiated oestrogen sulfates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E9505
β-Estradiol 3-sulfate sodium salt, ≥93%
C18H23NaO5S