EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
3-hydroxymethyl-beta-carboline antagonizes some pharmacologic actions of diazepam.
PMID 6265233
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E004
3-Hydroxymethyl-β-carboline
C12H10N2O