EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Inhibition of calmodulin-activated cyclic nucleotide phosphodiesterase by Triton X-100.
PMID 6268071
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
QS15 Triton QS-15