EMAIL THIS PAGE TO A FRIEND
Canadian journal of biochemistry
Hydrolysis of triolein, cholesterol oleate, and 4-methylumbelliferyl stearate by acid and neutral ester hydrolases (lipases) from pigeon adipose tissue: effect of cAMP-dependent protein kinase.
PMID 6271359