EMAIL THIS PAGE TO A FRIEND
Cytometry
Enzyme kinetic studies in cell populations using fluorogenic substrates and flow cytometric techniques.
PMID 6271514
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0883
4-Methylumbelliferyl acetate, esterase substrate
C12H10O4