EMAIL THIS PAGE TO A FRIEND
Acta chemica Scandinavica. Series B: Organic chemistry and biochemistry
Muscimol, a psychoactive constituent of Amanita muscaria, as a medicinal chemical model structure.
PMID 6274117
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1523
Muscimol, powder
C4H6N2O2