EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Inhibitors of specific aminoacyl-tRNA synthetases prevent thyrotropin-induced desensitization in cultured human thyroid cells.
PMID 6276382