EMAIL THIS PAGE TO A FRIEND
Revue d'oto-neuro-ophtalmologie
[Isolated sensory trigeminal neuropathy. Discussion on possible toxic etiology (author's transl)].
PMID 6278556
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185752
1-Chloronaphthalene, technical grade
C10H7Cl