EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Potential roles of the aminopyrimidine ring in thiamin catalyzed reactions.
PMID 6282162
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
261823
4-Aminopyrimidine, 98%
C4H5N3