EMAIL THIS PAGE TO A FRIEND
Mutation research
DNA repair studies on diallate, triallate and sulfallate in human cell cultures.
PMID 6283340
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45663
Sulfallate, PESTANAL®, analytical standard
C8H14ClNS2