EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
An electron spin resonance study of the novel radical cation produced during the horseradish peroxidase-catalyzed oxidation of tetramethylhydrazine.
PMID 6284143
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87860
Tetramethylhydrazine, ≥97.0%
C4H12N2