EMAIL THIS PAGE TO A FRIEND
Arzneimittel-Forschung
[On the site of action of benzarone and some other agents influencing the intracellular concentration of cyclic nucleotides].
PMID 6293514
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0490000 Benzarone, European Pharmacopoeia (EP) Reference Standard
C17H14O3