EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
A spin label substrate analogue as active site-directed modifying agent. Tryptophan 140 of aspartate aminotransferase.
PMID 6296136
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
220930
Potassium nitrosodisulfonate
K2NO7S2