EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur allgemeine Mikrobiologie
Immobilization of glucose isomerase.
PMID 6299015
Abstract
The immobilization of glucose isomerase (D-xylose ketol isomerase, EC 5.3.1.5) by covalently bonding to various carriers and by adsorption to ion exchange resins was attempted in order to obtain a stable immobilized enzyme which can be used for continuous isomerization of glucose in a column. Of the covalent bonding methods, the colloidal silica-glutaraldehyde method showed the highest binding capacity and gave the most stable immobilized glucose isomerase. The Ludox HS-30 bound glucose isomerase column showed a half-life of 24 days and an enzyme usage of 0.07 units per gram of isomerized sugar (d.s, fructose 45%). Of the resins used, the macromolecular type or porous type strongly basic anion exchange resins showed the highest binding capacity and gave the most stable immobilized glucose isomerase. The Amberlite IRA-904 resine-bound glucose isomerase showed a half-life of 23 days and an enzyme usage of 0.06 units per gram of isomerized sugar (d.s., fructose 45%). Based on the ease of the immobilization process, the possibility of carrier reuse and the extensive use already achieved by ion exchange resins in the sugar industry, IRA-904 resin was selected as the candidate for commercialization.