EMAIL THIS PAGE TO A FRIEND
The Journal of parasitology
Aminopeptidase activity in adult Schistosoma mansoni.
PMID 6304276
Abstract
An aminopeptidase activity capable of hydrolysing leucine 4-nitroanilide and alanine 4-nitroanilide at pH 7.0 was detected in saponin-CaCl2 extracts and homogenates of adult Schistosoma mansoni. The extracts were also capable of acting on synthetic dipeptides at the same pH, preferentially hydrolysing peptide bonds following leucine, alanine, or proline N-terminal residues. Imide bonds were not hydrolysed. The hydrolysis of leucine 4-nitroanilide was apparently stimulated by thiols, strongly inhibited by 1 mM 4-chloromercuric benzene sulfonic acid, and partially inhibited by 1 mM 1,10-phenanthroline.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9125
L-Leucine-p-nitroanilide, leucine aminopeptidase substrate
C12H17N3O3