EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Alterations in the activities of hepatic plasma-membrane and microsomal enzymes during liver regeneration.
PMID 6309144
Abstract
A marked increase in the activities of rat liver plasma-membrane (Na+ + K+)-stimulated ATPase and microsomal Ca2+-stimulated ATPase was observed 18h after partial hepatectomy. Lipid analyses for both membrane preparations reveal that in partially hepatectomized rats the cholesterol and sphingomyelin content are decreased with a subsequent decrease in the cholesterol/phospholipid molar ratio compared with those of sham-operated animals. Changes in the allosteric properties of plasma-membrane (Na+ + K+)-stimulated ATPase by F- (as reflected by changes in the Hill coefficient) indicated a fluidization of the lipid bilayer of both membrane preparations in 18 h-regenerating liver. The amphipathic dodecyl glucoside incorporated into the hepatic plasma membranes evoked a marked increase in the (Na+ + K+)-stimulated ATPase and 5'-nucleotidase activities. The lack of effect of the glucoside on the Lubrol-PX-solubilized 5'-nucleotidase indicates that changes in the activities of the membrane-bound enzymes caused by the glucoside are due to modulation of the membrane fluidity. Dodecyl glucoside appears to increase the membrane fluidity, evaluated through changes in the Hill coefficient for plasma-membrane (Na+ + K+)-stimulated ATPase. The biological significance of these data is discussed in terms of the differences and changes in the interaction of membrane-bound enzymes with membrane lipids during liver regeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8035
n-Dodecyl β-D-glucopyranoside, ≥98% (GC)
C18H36O6