EMAIL THIS PAGE TO A FRIEND
European heart journal
Effects of molsidomine and other NO-containing vasodilators on cyclic GMP formation.
PMID 6311553
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2901
Molsidomine
C9H14N4O4
Y0000821
Molsidomine, European Pharmacopoeia (EP) Reference Standard
C9H14N4O4