EMAIL THIS PAGE TO A FRIEND
Physiological reviews
Formation and actions of leukotrienes.
PMID 6324254
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5011
Leukotriene D4, ~50 μg/mL (in methanol/ammonium acetate buffer, 70:30, pH 5.6), ≥97%
C25H40N2O6S