EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Hygienic characteristics of the pesticide polychom].
PMID 6325304
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45577 Metiram, PESTANAL®, analytical standard