EMAIL THIS PAGE TO A FRIEND
Molecular and biochemical parasitology
Inhibition by superoxide dismutase and catalase of the damage of isolated Leishmania mexicana amazonensis by phenazine methosulfate.
PMID 6328296
Abstract
Phenazine methosulfate, a cationic electron carrier, inhibits the extracellular growth of promastigotes and the conversion of amastigotes into promastigote forms of Leishmania mexicana amazonensis. Growth inhibition and damage of extracellular parasites by PMS was counteracted by superoxide dismutase, a scavenger of the superoxide anion (O2-), and to a lesser extent, by catalase, a scavenger of hydrogen peroxide (H2O2). Inactivated dismutase and catalase were ineffective. Thus, damage of isolated L.m. amazonensis by phenazine methosulfate, involves the participation of O2- and H2O2. The role of the oxygen metabolites in the toxicity of phenazine methosulfate remains unknown. That O2- can damage the parasites is supported by the finding that superoxide dismutase also protected promastigotes from damage induced by oxygen intermediates generated by a xanthine-xanthine oxidase system. Killing of the parasites by crystal violet, a triphenylmethane, or basic blue 24, a phenothiazine, was not inhibited by superoxide dismutase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
556416
New Methylene Blue N, Dye content ~70 %
C18H22ClN3S