EMAIL THIS PAGE TO A FRIEND
Sbornik vedeckych praci Lekarske fakulty Karlovy univerzity v Hradci Kralove. Supplementum
[The efect of ionizing radiation and fasting on the excretion of o-iodobenzoic acid 125I and its conjugates in the urine of rats protected by radioprotective agents].
PMID 6335783
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
418463
2-Iodobenzoic acid, ≥99%, purified by sublimation
C7H5IO2
I7675
2-Iodobenzoic acid, 98%
C7H5IO2