EMAIL THIS PAGE TO A FRIEND
Experientia
Photobiological activity of 5,7-dimethoxycoumarin.
PMID 6337864
Abstract
Some biological properties of 5,7-dimethoxycoumarin (DMC) include dark induced frameshift mutagenesis in bacteria, lethal photosensitization and the formation of sister chromatid exchanges in Chinese hamster cells. The number of sister chromatid exchanges per unit of cell lethality produced by DMC is almost the same as observed with 5-methoxypsoralen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
116238
5,7-Dimethoxycoumarin, 98%
C11H10O4