EMAIL THIS PAGE TO A FRIEND
Mutation research
The biological activity of 4-chloromethylbiphenyl, benzyl chloride and 4-hydroxymethylbiphenyl in 4 short-term tests for carcinogenicity. A report of an individual study in the UKEMS genotoxicity trial 1981.
PMID 6338366
Abstract
4-Chloromethylbiphenyl (4CMB), benzyl chloride (BC) and 4-hydroxymethyl-biphenyl (4HMB) were tested for biological activity in the following assays: (i) the Salmonella/microsome assay; (ii) a bacterial 'fluctuation' assays; (iii) a DNA repair assay in Hela cells, and (iv) a mouse lymphoma mutation assay. 4CMB was active in assays (i), (ii) and (iii) but not in (iv); BC was active in assays (i), (ii), (iii) but not in (iv) while 4HMB was inactive in all assays. Where biological activity was seen this did not require addition of a liver S9 preparation. 4CMB was more active than BC in all the test systems in which a positive response was obtained. The implication of these results for a test battery approach to in vitro testing is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C52276
4-Phenylbenzyl chloride, 98%
C13H11Cl
123838
Biphenyl-4-methanol, 98%
C13H12O