EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Allylamine cardiotoxicity: metabolism and mechanism.
PMID 6346821
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
145831
Allylamine, 98%
C3H7N
241075
Allylamine, ≥99%
C3H7N