EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
DNA damage, prophage induction and mutation by furazolidone.
PMID 6349840
Abstract
Ultraviolet absorption data and thermal chromatography through hydroxyapatite (HAP) column revealed that furazolidone treatment of Vibrio cholerae cells produced more than 80% of DNA reversibly bihelical due to the formation of interstrand cross-links and the reaction obeyed a first order relation. Sensitivities of the Escherichia coli strains to the lethal action of the drug were in the order: AB 2480(uvr- rec-) greater than AB 2463(rec-) greater than AB 1886(uvr-) greater than AB 1157(repair proficient) or AB 4401(wild type). Furazolidone was 'Rec test' positive, produced dose-dependent prophage induction in E. coli cells and also dose-dependent streptomycin-resistance forward mutation in V. cholerae cells. The quantitative aspect and also the mode of furazolidone action on DNA were discussed.