EMAIL THIS PAGE TO A FRIEND
Pharmacotherapy
Pharmacology, clinical efficacy and adverse effects of vindesine sulfate, a new vinca alkaloid.
PMID 6359081
Abstract
Vindesine is a new vinca alkaloid antineoplastic agent derived from vinblastine. However, its antineoplastic spectrum more closely resembles that of vincristine. Clinical studies indicate activity against acute leukemia, lung cancer, carcinoma of the breast, squamous cell carcinoma of the esophagus and head and neck, Hodgkin's disease and non-Hodgkin's lymphomas. Pharmacokinetic studies indicate that vindesine exhibits a triphasic elimination pattern with a terminal half-life of 24.2 hours. Elimination is primarily through hepatic metabolism. The major side effects associated with vindesine therapy are myelosuppression and neurotoxicity. Other side effects include alopecia, nausea and vomiting and local tissue irritation associated with extravasation. Vindesine will be a positive addition to the antineoplastic armamentarium. The full extent of its activity remains to be established.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V0500000 Vindesine sulfate, European Pharmacopoeia (EP) Reference Standard
C43H55N5O7 · H2O4S
V4890
Vindesine sulfate salt hydrate, ≥95% (HPLC)
C43H55N5O7 · H2SO4 · xH2O