EMAIL THIS PAGE TO A FRIEND
Psychopharmacology bulletin
The efficacy of amoxapine, maprotiline, and trazodone in comparison to imipramine and amitriptyline: a review of the literature.
PMID 6374720
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A129
Amoxapine
C17H16ClN3O