EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Excretion of catecholamines and their metabolites in transplantable rat phaeochromocytoma.
PMID 6375678
Abstract
The urinary excretion pattern of catecholamines and their metabolites was studied in rats bearing a subcutaneous transplantable phaeochromocytoma. Compared with normal rats, tumour-bearing animals showed a markedly raised excretion of dopamine, noradrenaline and adrenaline, together with certain of their major acidic and alcoholic metabolites. No evidence of increased octopamine production could be obtained. There was a significant correlation between the output of dopamine and its metabolites, allowing accurate assessment of dopamine turnover rates which were comparable with those observed in human phaeochromocytoma. Tumour development, as determined by tumour weight, also correlated significantly with urinary excretion of noradrenaline and dopamine. Rat phaeochromocytoma appears to be a useful model for the human tumour.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
148830
Homovanillyl alcohol, 99%
C9H12O3