EMAIL THIS PAGE TO A FRIEND
CRC critical reviews in biochemistry
Thermodynamics of hapten-antibody interactions.
PMID 6375963
Abstract
Two schools of thought are currently prevalent regarding the thermodynamic mechanism(s) of hapten-antibody interaction(s). While one school is a proponent of the hapten-antibody reaction being driven predominantly by enthalpy, the second school rationalizes the mechanism as an enthalpy-entropy compensation, the magnitude of the latter being dependent on the temperature at which hapten-antibody interaction is carried out.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
200182
Nitrazine Yellow, indicator grade, Dye content 85 %
C16H8N4Na2O11S2