EMAIL THIS PAGE TO A FRIEND
Nihon Ika Daigaku zasshi
[Influences of oral hypoglycemic agents on the beta-cell of the rabbit pancreas].
PMID 6378959
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
392340
Potassium rhodizonate, technical grade, 85%
C6K2O6