EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Hautkrankheiten
[Efficacy and safety of topical 0.05% diflorasone diacetate (florone) cream/ointment in patients with acute/chronic dermatitis].
PMID 6391003
Abstract
In a three weeks study, efficacy and local tolerance of 0.05% diflorasone diacetate (Florone) cream and ointment were tested in 60 patients suffering from eczematous dermatosis or dermatitis. The results have been judged by 8 signs of improvement and are presented in a chart. Both formulations showed an efficacy of 95% during the 21-day duration of therapy. Side effects were rarely observed.