EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Photoaffinity reagents for use with pepsin and other carboxyl proteases.
PMID 6404274
Abstract
Two compounds have been designed to serve as photoaffinity reagents for use with carboxyl proteases. 1,2-Epoxy-3-(4'-azido-2'-nitrophenoxy)propane has been synthesized and shown to react with porcine pepsin in the same fashion as the traditional inhibitor 1,2-epoxy-3-(p-nitrophenoxy)propane, while p-azidophenacyl bromide is similar to other phenacyl bromides in its reaction with pepsin. In combination with p-azido-alpha-diazoacetophenone, previously shown to resemble alpha-diazo carbonyl reagents in its reaction with pepsin, photoaffinity analogs are now available for all three of the widely-used carboxyl protease inhibitors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6057
4-Azidophenacyl bromide, powder
C8H6BrN3O