EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Vitamin K biosynthesis in bacteria--precursors, intermediates, enzymes, and genes.
PMID 6406647
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
281255
1,4-Dihydroxy-2-naphthoic acid, 97%
C11H8O4