EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Late sequelae of the action of diphenyl derivatives].
PMID 6407901
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151378
Bis(4-chlorophenyl) sulfone, 98%
C12H8Cl2O2S