EMAIL THIS PAGE TO A FRIEND
MMW, Munchener medizinische Wochenschrift
[Benzarone for leg edema caused by chronic venous insufficiency].
PMID 6408456
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0490000 Benzarone, European Pharmacopoeia (EP) Reference Standard
C17H14O3