EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
A convener role for the cnxH gene specified component in the NADPH-nitrate reductase fron Aspergillus nidulans.
PMID 6423961
Abstract
The role of the cnxH+ gene specified polypeptide in the formation and function of the NADPH-nitrate reductase in Aspergillus nidulans was examined with the use of two complementing mutant strains which were grown as forced heterocaryons in the presence of nitrate. The niaD-421 structural gene mutant and the cnxH-318 co-factor gene mutant produce two components of the NADPH-cytochrome c reductase co-activity which can be distinguished by their enzymatic and physical behavior. This combination enabled us to isolate the de novo synthesis of niaD+ gene specified protomers from the constitutively formed co-factor at two stages of development. The proportion of induced and constitutively formed protomers in the isolated holoenzyme was measured after pulsing with [3H]-histidine or [14C]-histidine prior to induction with nitrate. The newly formed nitrate reductase was resolved by agarose gel electrofocusing and activity staining. In vivo assembly of a 7.8s enzyme in the heterocaryotic mycelium of the above strains is apparently achieved by the convener action of the cnxH+ gene directed polypeptide from the niaD- strain on the niaD+ gene directed protomers of the cnxH- partner. This occurs with or without Mo as a co-factor.
Related Materials