EMAIL THIS PAGE TO A FRIEND
The International journal of biochemistry
Circular dichroism study of thyroxine-binding globulin derivatized with p-iodophenylsulfonyl chloride.
PMID 6427036
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
222941
4-Iodobenzenesulfonyl chloride, 95%
C6H4ClIO2S