EMAIL THIS PAGE TO A FRIEND
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme
[DABITC microsequence analysis of proteins].
PMID 6431496
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
317802
4-(4-Isothiocyanatophenylazo)-N,N-dimethylaniline, 97%
C15H14N4S