Journal of biochemistry
Purification of several bacteriolytic enzymes by affinity chromatography on lysozyme-lysate of Micrococcus lysodeikticus cell wall coupled with sepharose.
PMID 6436
Using lysozyme-lysate of Micrococcus lysodeikticus cell wall coupled with Sepharose, several bacteriolytic enzymes were purified from crude preparations of animal and microbial origin. Quail egg-white, human milk and salivary lysozymes [EC] were adsorbed onto the adsorbent at pH 5-7 and eluted with 2M NaCl at pH 10. By means of these treatments, lysozymes were purified 20-250 fold with activity recoveries of 60-80%, and the quail lysozyme thus purified was shown to be discelectrophoretically homogeneous. Some bacteriolytic enzymes of microbial origin were also highly purified by using this affinity adsorbent. A bacterial lysozyme from Bacillus sp. ML-208 showed high affinity for the ligand and was not eluted under the conditions mentioned above, but was recovered by elution with 2M guanidine-HCl at pH 5.8, resulting in a 500-fold increase in the specific activity. A Pseudomonas-lytic enzyme from Streptomyces sp. P-51 was easily released from the adsorbent by elution with 0.5M NaCl at pH 5.0. A staphylolytic F2 enzyme from S. griseus S-35 and a chitinase [EC] from yam, both of which were completely inert toward M. lysodeikticus cell wall, passed through the adsorbent column. A modified ligand, in which muramic acid and glucosamine residues were N,O-acetylated, failed to adsorb any of these animal and bacterial lysozymes. Some of the enzymatic properties and bacteriolytic action spectra of these purified enzymes are also described in this paper in comparison with those of hen egg-white lysozyme.